Product Details

SNP ID
rs72789330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31436503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCTGGTACTGGGGACAGACGC[A/C]GGGCTTCGGCCAGTGTGTGCTCCTT
Phenotype
MIM: 602009 MIM: 602453
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
COX6A2 PubMed Links

Gene Details

Gene
COX6A2
Gene Name
cytochrome c oxidase subunit 6A2
There are no transcripts associated with this gene.

Gene
ITGAD
Gene Name
integrin subunit alpha D
There are no transcripts associated with this gene.

Gene
ZNF843
Gene Name
zinc finger protein 843
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136509.1 1074 Missense Mutation CGG,CTG R116L NP_001129981.1

View Full Product Details