Product Details

SNP ID
rs74006128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81905059 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCTCCGAGGTGCCCCCACCCA[A/G]CTGCACCTGTTGGTGATGATCCGCT
Phenotype
MIM: 614534 MIM: 607996 MIM: 602679 MIM: 606212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANAPC11 PubMed Links

Gene Details

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
There are no transcripts associated with this gene.

Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184917.2 Intron NP_001171846.1
NM_001256433.2 Intron NP_001243362.1
NM_001256434.2 Intron NP_001243363.1
NM_001256435.2 Intron NP_001243364.1
NM_001282203.1 Intron NP_001269132.1
NM_001282204.1 Intron NP_001269133.1
NM_002861.4 Intron NP_002852.1
XM_005256386.3 Intron XP_005256443.1
XM_005256387.3 Intron XP_005256444.1
XM_006722287.3 Intron XP_006722350.1
XM_017024910.1 Intron XP_016880399.1
XM_017024911.1 Intron XP_016880400.1
XM_017024912.1 Intron XP_016880401.1
Gene
SIRT7
Gene Name
sirtuin 7
There are no transcripts associated with this gene.

View Full Product Details