Product Details

SNP ID
rs71373406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41308342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGGTTGGCAGATACTGGAGAC[A/G]CAACAAGAAGGCTCTTGGCAAGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP16-1 PubMed Links

Gene Details

Gene
KRTAP16-1
Gene Name
keratin associated protein 16-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146182.1 912 Silent Mutation TGC,TGT C304C NP_001139654.1
Gene
KRTAP17-1
Gene Name
keratin associated protein 17-1
There are no transcripts associated with this gene.

Gene
KRTAP29-1
Gene Name
keratin associated protein 29-1
There are no transcripts associated with this gene.

View Full Product Details