Product Details

SNP ID
rs72821577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39067091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAACTGAGTCCAAATTCATTCT[G/T]CTTGCTGCGTCATAGCCAATACGTT
Phenotype
MIM: 606826
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100131347 PubMed Links

Gene Details

Gene
LOC100131347
Gene Name
RAD52 motif containing 1 pseudogene
There are no transcripts associated with this gene.

Gene
PLXDC1
Gene Name
plexin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020405.4 2464 UTR 3 NP_065138.2

View Full Product Details