Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002146.4 | 2017 | Intron | NP_002137.4 | ||
XM_005257277.3 | 2017 | Intron | XP_005257334.1 | ||
XM_005257280.3 | 2017 | Intron | XP_005257337.1 | ||
XM_005257282.3 | 2017 | Intron | XP_005257339.1 | ||
XM_006721854.2 | 2017 | Intron | XP_006721917.1 | ||
XM_011524707.2 | 2017 | Intron | XP_011523009.1 | ||
XM_011524708.2 | 2017 | Intron | XP_011523010.1 | ||
XM_011524710.1 | 2017 | Intron | XP_011523012.1 | ||
XM_011524719.1 | 2017 | Intron | XP_011523021.1 | ||
XM_011524720.2 | 2017 | Intron | XP_011523022.1 | ||
XM_011524721.2 | 2017 | Intron | XP_011523023.1 | ||
XM_011524726.2 | 2017 | Intron | XP_011523028.1 | ||
XM_017024557.1 | 2017 | Intron | XP_016880046.1 | ||
XM_017024558.1 | 2017 | Intron | XP_016880047.1 | ||
XM_017024559.1 | 2017 | Intron | XP_016880048.1 | ||
XM_017024560.1 | 2017 | Intron | XP_016880049.1 | ||
XM_017024561.1 | 2017 | Intron | XP_016880050.1 | ||
XM_017024562.1 | 2017 | Intron | XP_016880051.1 | ||
XM_017024563.1 | 2017 | Intron | XP_016880052.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_024015.4 | 2017 | UTR 3 | NP_076920.1 |