Product Details

SNP ID
rs72847554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28732501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGTAGAAATAGGATTTGCTTGT[A/G]TGAAATGGAGGAAAGATTCTAGGGC
Phenotype
MIM: 609799 MIM: 602326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEK8 PubMed Links

Gene Details

Gene
NEK8
Gene Name
NIMA related kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178170.2 Intron NP_835464.1
XM_011524638.2 Intron XP_011522940.1
XM_011524640.2 Intron XP_011522942.1
XM_017024499.1 Intron XP_016879988.1
XM_017024500.1 Intron XP_016879989.1
XM_017024501.1 Intron XP_016879990.1
Gene
RPL23A
Gene Name
ribosomal protein L23a
There are no transcripts associated with this gene.

Gene
SNORD42A
Gene Name
small nucleolar RNA, C/D box 42A
There are no transcripts associated with this gene.

Gene
SNORD4A
Gene Name
small nucleolar RNA, C/D box 4A
There are no transcripts associated with this gene.

Gene
SNORD4B
Gene Name
small nucleolar RNA, C/D box 4B
There are no transcripts associated with this gene.

Gene
TLCD1
Gene Name
TLC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details