Product Details

SNP ID
rs71353250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:13221218 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTTTGGAAATAGTTAACAGAA[C/T]GTATAGCGGCACATGGATATTAAAT
Phenotype
MIM: 606571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDLRAD4 PubMed Links

Gene Details

Gene
LDLRAD4
Gene Name
low density lipoprotein receptor class A domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003674.3 Intron NP_001003674.1
NM_001003675.3 Intron NP_001003675.1
NM_001276249.1 Intron NP_001263178.1
NM_001276251.1 Intron NP_001263180.1
NM_181481.4 Intron NP_852146.1
NM_181482.4 Intron NP_852147.1
XM_005258140.1 Intron XP_005258197.1
XM_006722353.1 Intron XP_006722416.1
XM_006722354.1 Intron XP_006722417.1
XM_011525737.1 Intron XP_011524039.1
XM_011525738.1 Intron XP_011524040.1
XM_017025961.1 Intron XP_016881450.1
XM_017025962.1 Intron XP_016881451.1
XM_017025963.1 Intron XP_016881452.1
XM_017025964.1 Intron XP_016881453.1
XM_017025965.1 Intron XP_016881454.1
XM_017025966.1 Intron XP_016881455.1
XM_017025967.1 Intron XP_016881456.1
XM_017025968.1 Intron XP_016881457.1
XM_017025969.1 Intron XP_016881458.1
XM_017025970.1 Intron XP_016881459.1
XM_017025971.1 Intron XP_016881460.1
XM_017025972.1 Intron XP_016881461.1

View Full Product Details