Product Details
- SNP ID
-
rs73930643
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39782607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTTGCTAGGAAAACATATTGGTG[C/T]CCAATGTCATGGAGAGGTTGATGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LEUTX
PubMed Links
Gene Details
- Gene
- LEUTX
- Gene Name
- leucine twenty homeobox
View Full Product Details