Product Details

SNP ID
rs73056666
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46616280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAATATACAGGAGGCTTAGAAGG[A/G]AGGCATCAGATGCCCCAAGTTGGAG
Phenotype
MIM: 114183 MIM: 600022
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CALM3 PubMed Links

Gene Details

Gene
CALM3
Gene Name
calmodulin 3
There are no transcripts associated with this gene.

Gene
PTGIR
Gene Name
prostaglandin I2 (prostacyclin) receptor (IP)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000960.3 Intron NP_000951.1
XM_005259093.2 Intron XP_005259150.1
XM_005259095.3 Intron XP_005259152.1

View Full Product Details