Product Details
- SNP ID
-
rs73932655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55486914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGGCCGAGGGGGGCTGGGGCTG[C/T]CTGGGCTACACGCTGGTGAGGGAAT
- Phenotype
-
MIM: 610671
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NAT14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77664073] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NAT14
- Gene Name
- N-acetyltransferase 14 (putative)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020378.3 |
696 |
Silent Mutation |
TGC,TGT |
C193C |
NP_065111.1 |
- Gene
- SSC5D
- Gene Name
- scavenger receptor cysteine rich family member with 5 domains
There are no transcripts associated with this gene.
- Gene
- ZNF628
- Gene Name
- zinc finger protein 628
There are no transcripts associated with this gene.
View Full Product Details