Product Details

SNP ID
rs73056477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57535822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTACCTTCCAGGACCCAGATAGT[A/C]TCCTGACTTAAGCTGGGGACAGGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF549 PubMed Links

Gene Details

Gene
ZNF549
Gene Name
zinc finger protein 549
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199295.1 Intron NP_001186224.1
NM_153263.2 Intron NP_694995.2
XM_006723128.3 Intron XP_006723191.1
Gene
ZNF550
Gene Name
zinc finger protein 550
There are no transcripts associated with this gene.

View Full Product Details