Product Details

SNP ID
rs73931236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35629120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCAGCGGCGGCGGCTAAGCAGA[A/G]ACTGTAGTAGCGGCGACAGCGACGA
Phenotype
MIM: 613432 MIM: 613232
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS5 PubMed Links

Gene Details

Gene
HAUS5
Gene Name
HAUS augmin like complex subunit 5
There are no transcripts associated with this gene.

Gene
RBM42
Gene Name
RNA binding motif protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319113.1 105 UTR 5 NP_001306042.1
NM_024321.4 105 UTR 5 NP_077297.2
XM_005259257.2 105 UTR 5 XP_005259314.1

View Full Product Details