Product Details
- SNP ID
-
rs73622721
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37150671 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATATGGTTGTGTTGGTTTTCAT[A/G]TAAGTTATTAATCTGTTACATGAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF585A
PubMed Links
Gene Details
- Gene
- ZNF585A
- Gene Name
- zinc finger protein 585A
View Full Product Details