Product Details

SNP ID
rs73539613
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5910179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGAACAAGGAGGGGGGTTGTT[C/T]GTACAAAGGAAAGGCAGGCTGGGGC
Phenotype
MIM: 114212 MIM: 612638 MIM: 603327
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPS PubMed Links

Gene Details

Gene
CAPS
Gene Name
calcyphosine
There are no transcripts associated with this gene.

Gene
NDUFA11
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
There are no transcripts associated with this gene.

Gene
RANBP3
Gene Name
RAN binding protein 3
There are no transcripts associated with this gene.

Gene
VMAC
Gene Name
vimentin-type intermediate filament associated coiled-coil protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017921.3 1597 UTR 3 NP_001017921.1

View Full Product Details