Product Details

SNP ID
rs73914831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57351826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCATTTCTTTCGTAGCAGTTA[A/C]CAGTCTCTGAAACTAAGTCTGTTTC
Phenotype
MIM: 603343 MIM: 605114
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MTRNR2L3 PubMed Links

Gene Details

Gene
MTRNR2L3
Gene Name
MT-RNR2-like 3
There are no transcripts associated with this gene.

Gene
RAE1
Gene Name
ribonucleic acid export 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015885.1 265 UTR 5 NP_001015885.1
NM_003610.3 265 Intron NP_003601.1
XM_005260582.2 265 Intron XP_005260639.2
XM_005260583.2 265 Intron XP_005260640.1
XM_011529087.2 265 Intron XP_011527389.1
XM_011529088.2 265 UTR 5 XP_011527390.1
XM_011529089.1 265 Intron XP_011527391.1
XM_017028108.1 265 Intron XP_016883597.1
Gene
SPO11
Gene Name
SPO11, initiator of meiotic double stranded breaks
There are no transcripts associated with this gene.

View Full Product Details