Product Details

SNP ID
rs73277065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:19216020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTATGTGCCAGGCAATGCGCTAT[G/T]CCCACCTTATAGGTATTGCTTTATT
Phenotype
MIM: 609839
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC24A3 PubMed Links

Gene Details

Gene
SLC24A3
Gene Name
solute carrier family 24 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020689.3 Intron NP_065740.2

View Full Product Details