Product Details

SNP ID
rs66483039
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:22581315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGACCCGAGACCTGGATTTCACC[A/G]TGTCAAGATTGGGAATGCTTTTTTT
Phenotype
MIM: 600288
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FOXA2 PubMed Links

Gene Details

Gene
FOXA2
Gene Name
forkhead box A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021784.4 2099 UTR 3 NP_068556.2
NM_153675.2 2099 UTR 3 NP_710141.1
Gene
LINC00261
Gene Name
long intergenic non-protein coding RNA 261
There are no transcripts associated with this gene.

Gene
LINC01384
Gene Name
long intergenic non-protein coding RNA 1384
There are no transcripts associated with this gene.

View Full Product Details