Product Details
- SNP ID
-
rs72470559
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:22581322 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGACCTGGATTTCACCGTGTCAA[G/T]ATTGGGAATGCTTTTTTTTTCTTTT
- Phenotype
-
MIM: 600288
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112021763] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXA2
- Gene Name
- forkhead box A2
- Gene
- LINC00261
- Gene Name
- long intergenic non-protein coding RNA 261
There are no transcripts associated with this gene.
- Gene
- LINC01384
- Gene Name
- long intergenic non-protein coding RNA 1384
There are no transcripts associated with this gene.
View Full Product Details