Product Details

SNP ID
rs73357389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34669339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTTGCCCAGCGCCACCGACCC[C/T]TAAGCAGCGTCAAGGAAGGAGTCCC
Phenotype
MIM: 615321
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLIC6 PubMed Links

Gene Details

Gene
CLIC6
Gene Name
chloride intracellular channel 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317009.1 387 UTR 5 NP_001303938.1
NM_053277.2 387 UTR 5 NP_444507.1
XM_017028406.1 387 UTR 5 XP_016883895.1

View Full Product Details