Product Details

SNP ID
rs73205541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38744480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCAAACTTCGCAAAGACCTGAA[C/T]AACTGAGGCAGCTAAGTGTTCGGGC
Phenotype
MIM: 611723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00114 PubMed Links

Gene Details

Gene
LINC00114
Gene Name
long intergenic non-protein coding RNA 114
There are no transcripts associated with this gene.

View Full Product Details