Product Details

SNP ID
rs73879002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23742571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGAGGGAGAGAAAGCCCTGTT[C/G]AATGAGAGGTCCCACTGCTCAGAAA
Phenotype
MIM: 194544
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF70 PubMed Links

Gene Details

Gene
ZNF70
Gene Name
zinc finger protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021916.3 3031 UTR 3 NP_068735.1

View Full Product Details