Product Details

SNP ID
rs73632955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2914819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAGAATTAGACCAAGTAATTGG[A/G]AATACTGAAGAAGCAAATAAATGAT
Phenotype
MIM: 300002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARSD PubMed Links

Gene Details

Gene
ARSD
Gene Name
arylsulfatase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001669.3 Intron NP_001660.2
NM_009589.3 Intron NP_033667.2
XM_005274514.2 Intron XP_005274571.1
XM_005274515.2 Intron XP_005274572.1

View Full Product Details