Product Details

SNP ID
rs72613040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19600799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTATTCCAATTCAGCATACAGGC[G/T]AAAATAGCAATTTTAAAGAGCAGAA
Phenotype
MIM: 400031
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TXLNGY PubMed Links

Gene Details

Gene
TXLNGY
Gene Name
taxilin gamma pseudogene, Y-linked
There are no transcripts associated with this gene.

View Full Product Details