Product Details

SNP ID
rs73238512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39048523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTATACTACATGCTGAAGCAG[A/G]TCAGCAAGAGAACTCCATTACAAAT
Phenotype
MIM: 608064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL5 PubMed Links

Gene Details

Gene
KLHL5
Gene Name
kelch like family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007075.2 Intron NP_001007076.1
NM_001171654.1 Intron NP_001165125.1
NM_015990.4 Intron NP_057074.3
NM_199039.3 Intron NP_950240.2
XM_005262655.4 Intron XP_005262712.1
XM_011513699.2 Intron XP_011512001.1
XM_011513700.2 Intron XP_011512002.1
XM_011513701.2 Intron XP_011512003.1
XM_017008273.1 Intron XP_016863762.1
XM_017008274.1 Intron XP_016863763.1
XM_017008275.1 Intron XP_016863764.1
XM_017008276.1 Intron XP_016863765.1

View Full Product Details