Product Details

SNP ID
rs73845733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121329662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGAAAATCCAATAATTTGCAC[A/G]ATAGCAAAAATCATCTTGATTGTGA
Phenotype
MIM: 606925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QRFPR PubMed Links

Gene Details

Gene
QRFPR
Gene Name
pyroglutamylated RFamide peptide receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198179.2 1327 Silent Mutation ATC,ATT I316I NP_937822.2
XM_017008693.1 1327 Intron XP_016864182.1

View Full Product Details