Product Details

SNP ID
rs73294632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48884467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACTAACTTCCCTTATCCTTA[C/T]ACATGCGCATAAACACACACACATG
Phenotype
MIM: 613316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC18 PubMed Links

Gene Details

Gene
LRRC18
Gene Name
leucine rich repeat containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006939.3 Intron NP_001006940.3
XM_006717867.2 Intron XP_006717930.1
XM_011539822.1 Intron XP_011538124.1
XM_011539823.1 Intron XP_011538125.1
XM_011539826.2 Intron XP_011538128.1
XM_011539828.2 Intron XP_011538130.1
XM_011539829.2 Intron XP_011538131.1
XM_017016276.1 Intron XP_016871765.1
XM_017016277.1 Intron XP_016871766.1
Gene
WDFY4
Gene Name
WDFY family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020945.1 Intron NP_065996.1
XM_005270004.3 Intron XP_005270061.1
XM_011539986.2 Intron XP_011538288.2
XM_011539987.2 Intron XP_011538289.2
XM_011539988.2 Intron XP_011538290.1
XM_011539990.2 Intron XP_011538292.1
XM_011539991.2 Intron XP_011538293.1
XM_011539992.2 Intron XP_011538294.1
XM_017016463.1 Intron XP_016871952.1
XM_017016464.1 Intron XP_016871953.1
XM_017016465.1 Intron XP_016871954.1

View Full Product Details