Product Details

SNP ID
rs72646588
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:109316863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTAACATACCTGTTTTTCCGGA[C/T]TCAAAAAGTTGCTTGTGCATTTCTT
Phenotype
MIM: 602458
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYBPHL PubMed Links

Gene Details

Gene
MYBPHL
Gene Name
myosin binding protein H like
There are no transcripts associated with this gene.

Gene
SORT1
Gene Name
sortilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205228.1 2404 Missense Mutation AAT,AGT N609S NP_001192157.1
NM_002959.6 2404 Missense Mutation AAT,AGT N746S NP_002950.3
XM_005271100.2 2404 Missense Mutation AAT,AGT N745S XP_005271157.1
XM_005271101.2 2404 Missense Mutation AAT,AGT N610S XP_005271158.1
XM_005271102.1 2404 Missense Mutation AAT,AGT N610S XP_005271159.1
XM_006710812.1 2404 Missense Mutation AAT,AGT N610S XP_006710875.1

View Full Product Details