Product Details

SNP ID
rs67289493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37085144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGGATAATGCTAAAGTATATT[C/T]TACCAATAGAAAGAGAGCTTTAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPATCH11 PubMed Links

Gene Details

Gene
GPATCH11
Gene Name
G-patch domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278505.1 Intron NP_001265434.1
NM_001322249.1 Intron NP_001309178.1
NM_174931.3 Intron NP_777591.3
XM_017003748.1 Intron XP_016859237.1
Gene
HEATR5B
Gene Name
HEAT repeat containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019024.2 Intron NP_061897.1
XM_006712034.2 Intron XP_006712097.1
XM_006712035.3 Intron XP_006712098.1
XM_011532934.2 Intron XP_011531236.1
XM_011532935.2 Intron XP_011531237.1
XM_017004378.1 Intron XP_016859867.1
XM_017004379.1 Intron XP_016859868.1
XM_017004380.1 Intron XP_016859869.1

View Full Product Details