Product Details

SNP ID
rs73928204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48687566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACTACACTTTCTACTAGGTAGA[A/G]GTCTCTTGCCTAATGTACCTAAAAA
Phenotype
MIM: 605358 MIM: 152790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2A1L PubMed Links

Gene Details

Gene
GTF2A1L
Gene Name
general transcription factor IIA subunit 1 like
There are no transcripts associated with this gene.

Gene
LHCGR
Gene Name
luteinizing hormone/choriogonadotropin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000233.3 2027 UTR 3 NP_000224.2
XM_005264309.3 2027 UTR 3 XP_005264366.1
XM_006712015.3 2027 UTR 3 XP_006712078.1
XM_011532828.1 2027 UTR 3 XP_011531130.1
XM_011532831.1 2027 UTR 3 XP_011531133.1
XM_011532834.2 2027 UTR 3 XP_011531136.1
XM_017004089.1 2027 UTR 3 XP_016859578.1
XM_017004090.1 2027 UTR 3 XP_016859579.1
Gene
STON1-GTF2A1L
Gene Name
STON1-GTF2A1L readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198593.1 2027 Intron NP_001185522.1
NM_001198594.1 2027 Intron NP_001185523.1
NM_172311.2 2027 Intron NP_758515.1

View Full Product Details