Product Details
- SNP ID
-
rs73149768
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:27373123 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAACAAAAGCCACAACACAAAAA[C/T]TAATCAGTGTGCAAAAATCTGATTA
- Phenotype
-
MIM: 603353
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NEK10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs67034824] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NEK10
- Gene Name
- NIMA related kinase 10
There are no transcripts associated with this gene.
- Gene
- SLC4A7
- Gene Name
- solute carrier family 4 member 7
View Full Product Details