Product Details

SNP ID
rs17080695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180847654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTATTGGAATTCCACTTTACCTC[A/G]CCACAAGGAGCTGGCTTTCATGACA
Phenotype
MIM: 610281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP62 PubMed Links

Gene Details

Gene
ZFP62
Gene Name
ZFP62 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172638.1 4011 UTR 3 NP_001166109.1
NM_152283.4 4011 UTR 3 NP_689496.4
XM_017009710.1 4011 UTR 3 XP_016865199.1
XM_017009711.1 4011 UTR 3 XP_016865200.1
XM_017009712.1 4011 UTR 3 XP_016865201.1
XM_017009713.1 4011 UTR 3 XP_016865202.1
XM_017009714.1 4011 UTR 3 XP_016865203.1
XM_017009715.1 4011 UTR 3 XP_016865204.1
XM_017009716.1 4011 Intron XP_016865205.1
XM_017009717.1 4011 UTR 3 XP_016865206.1
XM_017009718.1 4011 Intron XP_016865207.1
XM_017009719.1 4011 UTR 3 XP_016865208.1
XM_017009720.1 4011 UTR 3 XP_016865209.1
XM_017009721.1 4011 UTR 3 XP_016865210.1

View Full Product Details