Product Details

SNP ID
rs71558356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109224908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTTTGGAATGCCGATCTTTCT[A/G]TTTTAACGAACAAGAATGCACCATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100996634 PubMed Links

Gene Details

Gene
LOC100996634
Gene Name
transmembrane protein FLJ37396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277339.1 2354 Intron NP_001264268.1
XM_017010153.1 2354 UTR 5 XP_016865642.1
XM_017010154.1 2354 UTR 5 XP_016865643.1

View Full Product Details