Product Details

SNP ID
rs72728236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:65005202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTCCTGTGAGGTCAGGTATTTT[A/G]AAAAGGGAATTATAGGAAAAAATTT
Phenotype
MIM: 134636 MIM: 154950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHURC1-FNTB PubMed Links

Gene Details

Gene
CHURC1-FNTB
Gene Name
CHURC1-FNTB readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202558.1 Intron NP_001189487.1
NM_001202559.1 Intron NP_001189488.1
Gene
FNTB
Gene Name
farnesyltransferase, CAAX box, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002028.3 Intron NP_002019.1
Gene
MAX
Gene Name
MYC associated factor X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271068.1 Intron NP_001257997.1
NM_001271069.1 Intron NP_001257998.1
NM_001320415.1 Intron NP_001307344.1
NM_002382.4 Intron NP_002373.3
NM_145112.2 Intron NP_660087.1
NM_145113.2 Intron NP_660088.1
NM_145114.2 Intron NP_660089.1
NM_197957.3 Intron NP_932061.1
XM_011536773.2 Intron XP_011535075.1
XM_017021312.1 Intron XP_016876801.1
XM_017021313.1 Intron XP_016876802.1

View Full Product Details