Product Details

SNP ID
rs73358041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25120213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATTTAGTCTCTTCCTGATTTC[A/G]AAGAGCTCATTTACTATTTCGGGAA
Phenotype
MIM: 601491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPW PubMed Links

Gene Details

Gene
IPW
Gene Name
imprinted in Prader-Willi syndrome (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details