Product Details

SNP ID
rs72834380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:121741316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACACTTTGACTACTTGGATCAAT[A/G]CACTAGCTATTCTCAAACCAACATA
Phenotype
MIM: 607103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATE1 PubMed Links

Gene Details

Gene
ATE1
Gene Name
arginyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001976.2 3815 UTR 3 NP_001001976.1
NM_001288734.1 3815 UTR 3 NP_001275663.1
NM_001288735.1 3815 UTR 3 NP_001275664.1
NM_001288736.1 3815 UTR 3 NP_001275665.1
NM_007041.3 3815 UTR 3 NP_008972.2
XM_005269458.2 3815 UTR 3 XP_005269515.1
XM_005269459.3 3815 UTR 3 XP_005269516.1
XM_017015597.1 3815 UTR 3 XP_016871086.1
XM_017015598.1 3815 UTR 3 XP_016871087.1
XM_017015599.1 3815 UTR 3 XP_016871088.1
XM_017015600.1 3815 UTR 3 XP_016871089.1
XM_017015601.1 3815 Intron XP_016871090.1
XM_017015602.1 3815 UTR 3 XP_016871091.1
XM_017015603.1 3815 Intron XP_016871092.1
XM_017015604.1 3815 UTR 3 XP_016871093.1

View Full Product Details