Product Details

SNP ID
rs73214062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:20201529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAATTGTCTTTTTGTCCCCACT[C/T]GTAGTCCAAGTGCTAGAAGCTGTAT
Phenotype
MIM: 606939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1B2 PubMed Links

Gene Details

Gene
ATP6V1B2
Gene Name
ATPase H+ transporting V1 subunit B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001693.3 Intron NP_001684.2

View Full Product Details