Product Details

SNP ID
rs72963027
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:42177497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACTTAAGCAAACCATTGAAAA[C/T]CACCCCTCTTTTTTCAAGCATTTAC
Phenotype
MIM: 616035
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXJ3 PubMed Links

Gene Details

Gene
FOXJ3
Gene Name
forkhead box J3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198850.1 4298 UTR 3 NP_001185779.1
NM_001198851.1 4298 UTR 3 NP_001185780.1
NM_001198852.1 4298 UTR 3 NP_001185781.1
NM_014947.4 4298 UTR 3 NP_055762.3
XM_005270632.4 4298 UTR 3 XP_005270689.1
XM_006710458.3 4298 UTR 3 XP_006710521.1
XM_006710459.3 4298 UTR 3 XP_006710522.1
XM_011541026.2 4298 UTR 3 XP_011539328.1
XM_017000693.1 4298 UTR 3 XP_016856182.1
XM_017000694.1 4298 UTR 3 XP_016856183.1
XM_017000695.1 4298 UTR 3 XP_016856184.1

View Full Product Details