Product Details

SNP ID
rs72718703
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:84818598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTTAAAAAATAATGCTTAAAA[G/T]AATAAAAAAAATAAGAATACCTGTC
Phenotype
MIM: 605106
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LPAR3 PubMed Links

Gene Details

Gene
LPAR3
Gene Name
lysophosphatidic acid receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012152.2 Intron NP_036284.1

View Full Product Details