Product Details

SNP ID
rs73575411
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118897123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCGGCAGGATGCTGTTTCCCC[A/G]ATTCCATCCTCAGGGAGTGGAGACT
Phenotype
MIM: 609004 MIM: 601613
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BCL9L PubMed Links

Gene Details

Gene
BCL9L
Gene Name
B-cell CLL/lymphoma 9-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182557.2 6757 UTR 3 NP_872363.1
XM_005271511.2 6757 UTR 3 XP_005271568.1
XM_005271512.2 6757 Intron XP_005271569.1
XM_005271513.2 6757 Intron XP_005271570.1
XM_006718815.2 6757 UTR 3 XP_006718878.1
XM_011542756.2 6757 Intron XP_011541058.1
XM_017017572.1 6757 UTR 3 XP_016873061.1
XM_017017573.1 6757 Intron XP_016873062.1
XM_017017574.1 6757 Intron XP_016873063.1
Gene
CXCR5
Gene Name
C-X-C motif chemokine receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001716.4 6757 Intron NP_001707.1
NM_032966.2 6757 Intron NP_116743.1
Gene
LOC101929133
Gene Name
uncharacterized LOC101929133
There are no transcripts associated with this gene.

View Full Product Details