Product Details

SNP ID
rs73275932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43847423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACTGGACCAACCCAAGGCAAAA[C/T]TGTGATGCTGGTAGCACTTATTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00619 PubMed Links

Gene Details

Gene
LINC00619
Gene Name
long intergenic non-protein coding RNA 619
There are no transcripts associated with this gene.

View Full Product Details