Product Details

SNP ID
rs72776362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3381694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAACAACCATAGACAAATGTAC[C/T]TATCTTCTACCACTTTTTATCCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTRNR2L4 PubMed Links

Gene Details

Gene
MTRNR2L4
Gene Name
MT-RNR2-like 4
There are no transcripts associated with this gene.

Gene
ZSCAN32
Gene Name
zinc finger and SCAN domain containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284527.1 Intron NP_001271456.1
NM_001284528.1 Intron NP_001271457.1
NM_001284529.1 Intron NP_001271458.1
NM_001324340.1 Intron NP_001311269.1
NM_001324341.1 Intron NP_001311270.1
NM_001324342.1 Intron NP_001311271.1
NM_001324343.1 Intron NP_001311272.1
NM_001324344.1 Intron NP_001311273.1
NM_001324345.1 Intron NP_001311274.1
NM_001324346.1 Intron NP_001311275.1
NM_017810.3 Intron NP_060280.2
XM_005255403.2 Intron XP_005255460.1
XM_011522555.2 Intron XP_011520857.1
XM_017023371.1 Intron XP_016878860.1
XM_017023372.1 Intron XP_016878861.1
XM_017023373.1 Intron XP_016878862.1

View Full Product Details