Product Details

SNP ID
rs73491719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3382696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGCACAGCCAGGAGAGGTCAG[C/T]GTCCTTATGCTGACTCCTGGTCCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN32 PubMed Links

Gene Details

Gene
ZSCAN32
Gene Name
zinc finger and SCAN domain containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284527.1 2513 UTR 3 NP_001271456.1
NM_001284528.1 2513 UTR 3 NP_001271457.1
NM_001284529.1 2513 UTR 3 NP_001271458.1
NM_001324340.1 2513 UTR 3 NP_001311269.1
NM_001324341.1 2513 UTR 3 NP_001311270.1
NM_001324342.1 2513 UTR 3 NP_001311271.1
NM_001324343.1 2513 UTR 3 NP_001311272.1
NM_001324344.1 2513 UTR 3 NP_001311273.1
NM_001324345.1 2513 UTR 3 NP_001311274.1
NM_001324346.1 2513 UTR 3 NP_001311275.1
NM_017810.3 2513 UTR 3 NP_060280.2
XM_005255403.2 2513 UTR 3 XP_005255460.1
XM_011522555.2 2513 UTR 3 XP_011520857.1
XM_017023371.1 2513 UTR 3 XP_016878860.1
XM_017023372.1 2513 UTR 3 XP_016878861.1
XM_017023373.1 2513 UTR 3 XP_016878862.1

View Full Product Details