Product Details
- SNP ID
-
rs66593477
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70140730 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTGCCTTCACTGTAGTTTAATAT[C/T]GTTTAGGAGTTGAATACTGGACTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CSN1S2BP
PubMed Links
Gene Details
- Gene
- CSN1S2BP
- Gene Name
- casein alpha s2-like B, pseudogene
There are no transcripts associated with this gene.
View Full Product Details