Product Details

SNP ID
rs17139579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78110782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAATTTTTGAGAATTTTCATTTG[C/T]TTTTGCTTCCAAAAAAACAAACAGG
Phenotype
MIM: 311800
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGK1 PubMed Links

Gene Details

Gene
PGK1
Gene Name
phosphoglycerate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000291.3 Intron NP_000282.1

View Full Product Details