Product Details

SNP ID
rs78184329
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:161751936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATACGAAGTGGATACCTCTAGT[G/T]CAATTTATAAGCAATATCGTTTACA
Phenotype
MIM: 604835
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 563 Missense Mutation GCA,TCA A177S NP_009171.1
XM_005244862.2 563 Missense Mutation GCA,TCA A47S XP_005244919.1

View Full Product Details