Product Details

SNP ID
rs74743630
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:150730815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGCATCTATTTTCATTATGCT[C/T]ATCACTATTTCAAATTTAGGATAGC
Phenotype
MIM: 116845 MIM: 609824
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSS PubMed Links

Gene Details

Gene
CTSS
Gene Name
cathepsin S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199739.1 3488 UTR 3 NP_001186668.1
NM_004079.4 3488 UTR 3 NP_004070.3
Gene
HORMAD1
Gene Name
HORMA domain containing 1
There are no transcripts associated with this gene.

View Full Product Details