Product Details

SNP ID
rs75694492
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:151089907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTATACTTAGTACTTATTCAAT[G/T]GGTGGTATATTTATTTGTCCATTAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABPB2 PubMed Links

Gene Details

Gene
GABPB2
Gene Name
GA binding protein transcription factor beta subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323906.1 Intron NP_001310835.1
NM_001323907.1 Intron NP_001310836.1
NM_001323908.1 Intron NP_001310837.1
NM_001323909.1 Intron NP_001310838.1
NM_001323910.1 Intron NP_001310839.1
NM_001323911.1 Intron NP_001310840.1
NM_001323912.1 Intron NP_001310841.1
NM_001323913.1 Intron NP_001310842.1
NM_144618.2 Intron NP_653219.1
XM_017000246.1 Intron XP_016855735.1
XM_017000247.1 Intron XP_016855736.1
XM_017000248.1 Intron XP_016855737.1
XM_017000249.1 Intron XP_016855738.1
XM_017000250.1 Intron XP_016855739.1
XM_017000251.1 Intron XP_016855740.1
XM_017000252.1 Intron XP_016855741.1
XM_017000253.1 Intron XP_016855742.1
XM_017000254.1 Intron XP_016855743.1

View Full Product Details