Product Details

SNP ID
rs79670335
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:179302724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATGTCTCTAAGAAACCGGCTG[G/T]CAAAGTCCAGGGAAAATCCTGAGGA
Phenotype
MIM: 102642
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
SOAT1 PubMed Links

Gene Details

Gene
SOAT1
Gene Name
sterol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252511.1 271 UTR 5 NP_001239440.1
NM_001252512.1 271 Intron NP_001239441.1
NM_003101.5 271 Missense Mutation GCA,TCA A14S NP_003092.4
XM_011509911.1 271 Missense Mutation GCA,TCA A14S XP_011508213.1
XM_011509912.1 271 Missense Mutation GCA,TCA A14S XP_011508214.1

View Full Product Details