Product Details

SNP ID
rs76394976
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:185296608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACAGATGAGAGAAATAAAGTGG[C/T]AGAGTACCAGTGAACATTTGGAGAA
Phenotype
MIM: 609209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IVNS1ABP PubMed Links

Gene Details

Gene
IVNS1ABP
Gene Name
influenza virus NS1A binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006469.4 3844 UTR 3 NP_006460.2
XM_005244843.3 3844 UTR 3 XP_005244900.1
XM_011509079.2 3844 UTR 3 XP_011507381.1
XM_011509080.2 3844 UTR 3 XP_011507382.1
XM_017000084.1 3844 UTR 3 XP_016855573.1
Gene
SWT1
Gene Name
SWT1, RNA endoribonuclease homolog
There are no transcripts associated with this gene.

View Full Product Details