Product Details

SNP ID
rs77555743
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:153930009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCAATGCAGATCTGTGGGTAC[C/G]CTGGAGGGGAGTTCCCGGTATAGGA
Phenotype
MIM: 614998
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DENND4B PubMed Links

Gene Details

Gene
DENND4B
Gene Name
DENN domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014856.2 5337 UTR 3 NP_055671.2
XM_005245678.4 5337 UTR 3 XP_005245735.1
XM_005245679.2 5337 UTR 3 XP_005245736.1
XM_011510219.1 5337 UTR 3 XP_011508521.1
XM_011510220.2 5337 UTR 3 XP_011508522.1
XM_011510221.2 5337 UTR 3 XP_011508523.1
XM_011510222.1 5337 Intron XP_011508524.1
XM_017002993.1 5337 UTR 3 XP_016858482.1
Gene
GATAD2B
Gene Name
GATA zinc finger domain containing 2B
There are no transcripts associated with this gene.

View Full Product Details